accessibility menu, dialog, popup

UserName

Viewing profile as user:
NOHNNS

CAS RN 910037-26-2

IUPAC Name: (1-{thieno[3,2-d]pyrimidin-4-yl}piperidin-4-yl)methanol
Synonyms: (1-{thieno[3,2-d]pyrimidin-4-yl}piperidin-4-yl)methanol
Molecular Weight (g/mol): 249.33
Molecular Formula: C12H15N3OS
InChi Key: UXMVQNYPOJJGNA-UHFFFAOYSA-N
SMILES: OCC1CCN(CC1)C1=C2SC=CC2=NC=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results