CAS RN 910037-27-3
IUPAC Name:
ethyl 1-{thieno[3,2-d]pyrimidin-4-yl}piperidine-4-carboxylate
Synonyms:
ethyl 1-{thieno[3,2-d]pyrimidin-4-yl}piperidine-4-carboxylate
Molecular Weight (g/mol):
291.37
Molecular Formula:
C14H17N3O2S
InChi Key:
CBEGYEOZJHWLKV-UHFFFAOYSA-N
SMILES:
CCOC(=O)C1CCN(CC1)C1=C2SC=CC2=NC=N1
1
–
1
of
1
results
1
Ethyl 1-thieno[3,2-d]pyrimidin-4-ylpiperidine-4-carboxylate, 97%, Thermo Scientific™
CAS: 910037-27-3 Molecular Formula: C14H17N3O2S Molecular Weight (g/mol): 291.369 MDL Number: MFCD09065025 InChI Key: CBEGYEOZJHWLKV-UHFFFAOYSA-N Synonym: ethyl 1-thieno 3,2-d pyrimidin-4-yl piperidine-4-carboxylate,ethyl 1-thieno 3,2-d pyrimidin-4-ylpiperidine-4-carboxylate,4-piperidinecarboxylicacid,1-thieno 3,2-d pyrimidin-4-yl-,ethyl ester PubChem CID: 24229709 IUPAC Name: ethyl 1-thieno[3,2-d]pyrimidin-4-ylpiperidine-4-carboxylate SMILES: CCOC(=O)C1CCN(CC1)C2=NC=NC3=C2SC=C3