accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 910251-10-4

BOCH3CH3H3CFFFK

CAS RN 910251-10-4

IUPAC Name: potassium [(tert-butoxy)methyl]trifluoroboranuide
Synonyms: potassium (tert-butoxymethyl)trifluoroboranuide
Molecular Weight (g/mol): 194.05
Molecular Formula: C5H11BF3KO
InChi Key: XBEPCFNGTDJTIY-UHFFFAOYSA-N
SMILES: [K+].CC(C)(C)OC[B-](F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results