accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOOOHN

CAS RN 91092-92-1

IUPAC Name: methyl 1,4-dioxo-1,2,3,4-tetrahydroisoquinoline-3-carboxylate
Synonyms: methyl 1,4-dioxo-2,3-dihydroisoquinoline-3-carboxylate
Molecular Weight (g/mol): 219.2
Molecular Formula: C11H9NO4
InChi Key: FQCQGFASLAKUTO-UHFFFAOYNA-N
SMILES: COC(=O)C1NC(=O)C2=CC=CC=C2C1=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results