accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COBHOOH

CAS RN 91129-69-0

IUPAC Name: (2-butoxyphenyl)boronic acid
Synonyms: 2-butoxyphenylboronic acid
Molecular Weight (g/mol): 194.04
Molecular Formula: C10H15BO3
InChi Key: NNZPYUBZXKOFHS-UHFFFAOYSA-N
SMILES: CCCCOC1=CC=CC=C1B(O)O
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results