accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 91137-55-2

IUPAC Name: 4-methyl-2-phenyl-1,3-oxazole-5-carboxylic acid
Synonyms: 4-methyl-2-phenyl-1,3-oxazole-5-carboxylic acid
Molecular Weight (g/mol): 203.2
Molecular Formula: C11H9NO3
InChi Key: HRFYZRHGBICKAG-UHFFFAOYSA-N
SMILES: CC1=C(OC(=N1)C1=CC=CC=C1)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results