accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 911482-75-2

IUPAC Name: 5,5-dimethyl-2-(prop-2-en-1-yl)-1,3,2-dioxaborinane
Synonyms: 5,5-dimethyl-2-(prop-2-en-1-yl)-1,3,2-dioxaborinane
Molecular Weight (g/mol): 154.02
Molecular Formula: C8H15BO2
InChi Key: SBZNRMJWIVCYBF-UHFFFAOYSA-N
SMILES: CC1(C)COB(CC=C)OC1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results