accessibility menu, dialog, popup

UserName

Viewing profile as user:
BFFClHOOH

CAS RN 911645-24-4

IUPAC Name: (5-chloro-2,4-difluorophenyl)boronic acid
Synonyms: 5-chloro-2,4-difluorophenylboronic acid
Molecular Weight (g/mol): 192.35
Molecular Formula: C6H4BClF2O2
InChi Key: NEZHJPAWXWRHBO-UHFFFAOYSA-N
SMILES: OB(O)C1=CC(Cl)=C(F)C=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results