accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 91166-50-6

IUPAC Name: 2,3,3,6,6,7-hexamethyl-2,7-diaza-3,6-disilaoctane
Synonyms: 2,3,3,6,6,7-hexamethyl-2,7-diaza-3,6-disilaoctane
Molecular Weight (g/mol): 232.52
Molecular Formula: C10H28N2Si2
InChi Key: MRAAXSSHMOFDJR-UHFFFAOYSA-N
SMILES: CN(C)[Si](C)(C)CC[Si](C)(C)N(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results