accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 91183-71-0

IUPAC Name: tert-butyl (2S)-2-amino-4-(methylsulfanyl)butanoate hydrochloride
Synonyms: tert-butyl (2S)-2-amino-4-(methylsulfanyl)butanoate hydrochloride|H-MET-OTBU HCL
Molecular Weight (g/mol): 241.77
Molecular Formula: C9H20ClNO2S
InChi Key: KZJQROCWHZGZBJ-FJXQXJEOSA-N
SMILES: Cl.CSCC[C@H](N)C(=O)OC(C)(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)