accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CH3CCH3OONHNH

CAS RN 91188-13-5

IUPAC Name: tert-butyl N-(azetidin-3-yl)carbamate
Synonyms: tert-butyl N-(azetidin-3-yl)carbamate
Molecular Weight (g/mol): 172.23
Molecular Formula: C8H16N2O2
InChi Key: NEMXVXVJGXZDRR-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)NC1CNC1
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results