accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 91188-15-7

IUPAC Name: tert-butyl N-[(azetidin-3-yl)methyl]carbamate
Synonyms: tert-butyl N-(azetidin-3-ylmethyl)carbamate
Molecular Weight (g/mol): 186.26
Molecular Formula: C9H18N2O2
InChi Key: MOLUHRBHXXGWDP-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)NCC1CNC1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)