accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 912569-56-3

IUPAC Name: {3-[3-(dimethylamino)propoxy]phenyl}methanol
Synonyms: {3-[3-(dimethylamino)propoxy]phenyl}methanol
Molecular Weight (g/mol): 209.29
Molecular Formula: C12H19NO2
InChi Key: BABQTNDHVRVEJD-UHFFFAOYSA-N
SMILES: CN(C)CCCOC1=CC=CC(CO)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results