accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 912569-73-4

IUPAC Name: methyl({[1-methyl-5-phenoxy-3-(trifluoromethyl)-1H-pyrazol-4-yl]methyl})amine
Synonyms: methyl({[1-methyl-5-phenoxy-3-(trifluoromethyl)pyrazol-4-yl]methyl})amine
Molecular Weight (g/mol): 285.27
Molecular Formula: C13H14F3N3O
InChi Key: AJQKRVBDVZVOBK-UHFFFAOYSA-N
SMILES: CNCC1=C(OC2=CC=CC=C2)N(C)N=C1C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results