accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 912824-85-2

IUPAC Name: 4,4,5,5-tetramethyl-2-{8-oxatricyclo[7.4.0.0²,⁷]trideca-1(9),2(7),3,5,10,12-hexaen-6-yl}-1,3,2-dioxaborolane
Synonyms: 4,4,5,5-tetramethyl-2-{8-oxatricyclo[7.4.0.0²,⁷]trideca-1(9),2(7),3,5,10,12-hexaen-6-yl}-1,3,2-dioxaborolane
Molecular Weight (g/mol): 294.16
Molecular Formula: C18H19BO3
InChi Key: AVWYETCDUMCCNK-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=CC=CC2=C1OC1=C2C=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results