accessibility menu, dialog, popup

UserName

Viewing profile as user:
OOOHONHN(S)

CAS RN 913253-24-4

IUPAC Name: (2S)-4-cyano-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)butanoic acid
Synonyms: (2S)-4-cyano-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}butanoic acid
Molecular Weight (g/mol): 350.37
Molecular Formula: C20H18N2O4
InChi Key: IYSKJCGILKSAFL-SFHVURJKSA-N
SMILES: OC(=O)[C@H](CCC#N)NC(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2
Grade 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)