accessibility menu, dialog, popup

UserName

Viewing profile as user:
SOOOOOOHONHNNH2NH3CCH3CH3H3CCH3(S)CH3(E)

CAS RN 913733-27-4

IUPAC Name: (2S)-5-[(E)-[amino(2,2,4,6,7-pentamethyl-2,3-dihydro-1-benzofuran-5-sulfonamido)methylidene]amino]-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}(methyl)amino)pentanoic acid
Synonyms: (2S)-5-[(E)-[amino(2,2,4,6,7-pentamethyl-3H-1-benzofuran-5-sulfonamido)methylidene]amino]-2-{[(9H-fluoren-9-ylmethoxy)carbonyl](methyl)amino}pentanoic acid
Molecular Weight (g/mol): 662.8
Molecular Formula: C35H42N4O7S
InChi Key: MEGKXARLCPKZHJ-LJAQVGFWSA-N
SMILES: CN([C@@H](CCC\N=C(/N)NS(=O)(=O)C1=C(C)C2=C(OC(C)(C)C2)C(C)=C1C)C(O)=O)C(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2
Grade 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)