accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNNNH2H2OHClHClHCl

CAS RN 913830-33-8

IUPAC Name: 4-(4-methyl-1,4-diazepan-1-yl)aniline hydrate trihydrochloride
Synonyms: 4-(4-methyl-1,4-diazepan-1-yl)aniline hydrate trihydrochloride
Molecular Weight (g/mol): 332.69
Molecular Formula: C12H24Cl3N3O
InChi Key: RSGOMRQTIZQMIR-UHFFFAOYSA-N
SMILES: O.Cl.Cl.Cl.CN1CCCN(CC1)C1=CC=C(N)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results