accessibility menu, dialog, popup

UserName

Viewing profile as user:
BNNH3CCH3HOOH

CAS RN 913835-27-5

IUPAC Name: [4-(propan-2-yl)pyrimidin-5-yl]boronic acid
Synonyms: 4-isopropylpyrimidin-5-ylboronic acid
Molecular Weight (g/mol): 165.99
Molecular Formula: C7H11BN2O2
InChi Key: SMZXLPHORPDPKW-UHFFFAOYSA-N
SMILES: CC(C)C1=NC=NC=C1B(O)O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results