accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 913835-44-6

IUPAC Name: {4-[4-(2-hydroxyethyl)piperazine-1-carbonyl]phenyl}boronic acid hydrochloride
Synonyms: 4-[4-(2-hydroxyethyl)piperazine-1-carbonyl]phenylboronic acid hydrochloride
Molecular Weight (g/mol): 314.57
Molecular Formula: C13H20BClN2O4
InChi Key: VZGUMMZRYDMYMP-UHFFFAOYSA-N
SMILES: Cl.OCCN1CCN(CC1)C(=O)C1=CC=C(C=C1)B(O)O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results