accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 913835-52-6

IUPAC Name: [4-(acetamidosulfonyl)phenyl]boronic acid
Synonyms: 4-(acetamidosulfonyl)phenylboronic acid
Molecular Weight (g/mol): 243.04
Molecular Formula: C8H10BNO5S
InChi Key: CEUUKZMDDOCDTD-UHFFFAOYSA-N
SMILES: CC(=O)NS(=O)(=O)C1=CC=C(C=C1)B(O)O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results