accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 913835-67-3

BNOOH3CH3CCH3NHOHO

CAS RN 913835-67-3

IUPAC Name: {1-[(tert-butoxy)carbonyl]-6-cyano-1H-indol-2-yl}boronic acid
Synonyms: 1-(tert-butoxycarbonyl)-6-cyanoindol-2-ylboronic acid
Molecular Weight (g/mol): 286.09
Molecular Formula: C14H15BN2O4
InChi Key: CZEBTZMIQZVUSE-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)N1C(=CC2=CC=C(C=C12)C#N)B(O)O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results