accessibility menu, dialog, popup

UserName

Viewing profile as user:
BOOHClHOOH

CAS RN 913835-75-3

IUPAC Name: 4-chloro-3-(dihydroxyboranyl)benzoic acid
Synonyms: 4-chloro-3-(dihydroxyboranyl)benzoic acid
Molecular Weight (g/mol): 200.38
Molecular Formula: C7H6BClO4
InChi Key: ZITWKFSVFMUWIE-UHFFFAOYSA-N
SMILES: OB(O)C1=C(Cl)C=CC(=C1)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results