accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 913835-98-0

IUPAC Name: [6-(hydroxymethyl)pyridin-3-yl]boronic acid
Synonyms: 6-(hydroxymethyl)pyridin-3-ylboronic acid
Molecular Weight (g/mol): 152.94
Molecular Formula: C6H8BNO3
InChi Key: MSUQKXAJTUAESD-UHFFFAOYSA-N
SMILES: OCC1=NC=C(C=C1)B(O)O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results