accessibility menu, dialog, popup

UserName

Viewing profile as user:
BOOHOCH3HOOH

CAS RN 913836-10-9

IUPAC Name: 3-(dihydroxyboranyl)-2-methoxybenzoic acid
Synonyms: 3-(dihydroxyboranyl)-2-methoxybenzoic acid
Molecular Weight (g/mol): 195.97
Molecular Formula: C8H9BO5
InChi Key: QUWLFFYGKXZCER-UHFFFAOYSA-N
SMILES: COC1=C(C=CC=C1C(O)=O)B(O)O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results