CAS RN 914399-96-5
IUPAC Name:
3-(tert-butoxy)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-2-methylpropanoic acid
Synonyms:
3-(tert-butoxy)-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}-2-methylpropanoic acid
Molecular Weight (g/mol):
397.47
Molecular Formula:
C23H27NO5
InChi Key:
WRDFKKLMQLSBQN-HSZRJFAPSA-N
SMILES:
CC(C)(C)OCC(C)(NC(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2)C(O)=O
1
–
1
of
1
results
1
Fmoc-a-Me-D-Ser tBu -OH, Advanced ChemTech
CAS: 914399-96-5 Molecular Formula: C23H27NO5 Molecular Weight (g/mol): 397.47 MDL Number: MFCD26793608 InChI Key: WRDFKKLMQLSBQN-HSZRJFAPSA-N IUPAC Name: 3-(tert-butoxy)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-2-methylpropanoic acid SMILES: CC(C)(C)OCC(C)(NC(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2)C(O)=O