accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 914399-98-7

IUPAC Name: 3-(tert-butoxy)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-2-methylpropanoic acid
Synonyms: 3-(tert-butoxy)-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}-2-methylpropanoic acid
Molecular Weight (g/mol): 397.47
Molecular Formula: C23H27NO5
InChi Key: WRDFKKLMQLSBQN-QHCPKHFHSA-N
SMILES: CC(C)(C)OCC(C)(NC(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2)C(O)=O
Grade 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)