accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 914675-52-8

IUPAC Name: 2-{[1,1'-biphenyl]-2-yl}-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Synonyms: 2-{[1,1'-biphenyl]-2-yl}-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Molecular Weight (g/mol): 280.17
Molecular Formula: C18H21BO2
InChi Key: WCXWQEUBHZKNMQ-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=CC=CC=C1C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results