accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 915-05-9

IUPAC Name: (1R,3aS,3bS,7S,9aR,9bS,11aR)-1-[(2R,5R)-5-ethyl-6-methylheptan-2-yl]-9a,11a-dimethyl-1H,2H,3H,3aH,3bH,4H,6H,7H,8H,9H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-7-yl acetate
Synonyms: β-sitosteryl acetate
Molecular Weight (g/mol): 456.76
Molecular Formula: C31H52O2
InChi Key: PBWOIPCULUXTNY-LBKBYZTLSA-N
SMILES: CC[C@H](CC[C@@H](C)[C@H]1CC[C@H]2[C@@H]3CC=C4C[C@H](CC[C@]4(C)[C@H]3CC[C@]12C)OC(C)=O)C(C)C
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results