accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOH3COH3CH3COCH3O

CAS RN 915-35-5

IUPAC Name: 5,7',9',13'-tetramethyl-10'-oxo-5'-oxaspiro[oxane-2,6'-pentacyclo[10.8.0.0²,⁹.0⁴,⁸.0¹³,¹⁸]icosan]-16'-yl acetate
Synonyms: 5,7',9',13'-tetramethyl-10'-oxo-5'-oxaspiro[oxane-2,6'-pentacyclo[10.8.0.0²,⁹.0⁴,⁸.0¹³,¹⁸]icosan]-16'-yl acetate
Molecular Weight (g/mol): 472.67
Molecular Formula: C29H44O5
InChi Key: CVKZWRTYHCDWTE-UHFFFAOYNA-N
SMILES: CC1C2C(CC3C4CCC5CC(CCC5(C)C4CC(=O)C23C)OC(C)=O)OC11CCC(C)CO1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results