accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 915402-03-8

IUPAC Name: trimethyl({2-[3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]ethynyl})silane
Synonyms: trimethyl({2-[3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]ethynyl})silane
Molecular Weight (g/mol): 300.28
Molecular Formula: C17H25BO2Si
InChi Key: LDKJXCOLGXLWKP-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=CC(=CC=C1)C#C[Si](C)(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results