accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NNOHO

CAS RN 915707-39-0

IUPAC Name: 4-(1-methyl-1H-pyrazol-3-yl)benzoic acid
Synonyms: 4-(1-methylpyrazol-3-yl)benzoic acid
Molecular Weight (g/mol): 202.21
Molecular Formula: C11H10N2O2
InChi Key: ZIRNSRSOPMSFDK-UHFFFAOYSA-N
SMILES: CN1C=CC(=N1)C1=CC=C(C=C1)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results