accessibility menu, dialog, popup

UserName

Viewing profile as user:
LiCH3NNOO

CAS RN 915707-44-7

IUPAC Name: lithium(1+) 2-[(4-methylpiperazin-1-yl)methyl]benzoate
Synonyms: lithium(1+) 2-[(4-methylpiperazin-1-yl)methyl]benzoate
Molecular Weight (g/mol): 240.23
Molecular Formula: C13H17LiN2O2
InChi Key: BHMRBFXQIGCNNV-UHFFFAOYSA-M
SMILES: [Li+].CN1CCN(CC2=CC=CC=C2C([O-])=O)CC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results