accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 915707-58-3

IUPAC Name: 4-methyl-2H,3H,4H-pyrido[3,2-b][1,4]oxazine-7-carboxylic acid
Synonyms: 4-methyl-2H,3H-pyrido[3,2-b][1,4]oxazine-7-carboxylic acid
Molecular Weight (g/mol): 194.19
Molecular Formula: C9H10N2O3
InChi Key: JTDUYLQIBHWRRK-UHFFFAOYSA-N
SMILES: CN1CCOC2=C1N=CC(=C2)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results