accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 915707-63-0

IUPAC Name: 4-[(6-methylpyrazin-2-yl)oxy]aniline
Synonyms: 4-[(6-methylpyrazin-2-yl)oxy]aniline
Molecular Weight (g/mol): 201.23
Molecular Formula: C11H11N3O
InChi Key: OZPUGVATAOJVLW-UHFFFAOYSA-N
SMILES: CC1=CN=CC(OC2=CC=C(N)C=C2)=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results