accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 915707-65-2

IUPAC Name: (4-bromo-1-methyl-1H-pyrazol-3-yl)methanol
Synonyms: (4-bromo-1-methylpyrazol-3-yl)methanol
Molecular Weight (g/mol): 191.03
Molecular Formula: C5H7BrN2O
InChi Key: JFGLJTFTVBZOCB-UHFFFAOYSA-N
SMILES: CN1C=C(Br)C(CO)=N1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results