accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 915707-66-3

IUPAC Name: 4-bromo-3-(chloromethyl)-1-methyl-1H-pyrazole
Synonyms: 4-bromo-3-(chloromethyl)-1-methylpyrazole
Molecular Weight (g/mol): 209.47
Molecular Formula: C5H6BrClN2
InChi Key: WHHNKVUVLZLAAM-UHFFFAOYSA-N
SMILES: CN1C=C(Br)C(CCl)=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results