accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHOHI

CAS RN 915707-73-2

IUPAC Name: 3-(hydroxymethyl)-4-iodophenol
Synonyms: 3-(hydroxymethyl)-4-iodophenol
Molecular Weight (g/mol): 250.04
Molecular Formula: C7H7IO2
InChi Key: BXWYBXMOICALJX-UHFFFAOYSA-N
SMILES: OCC1=C(I)C=CC(O)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results