accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClCH3OOCH3NH(R)NOOOO(R)NCH3H

CAS RN 91599-74-5

IUPAC Name: hydrogen 3-(3R)-1-benzylpiperidin-3-yl 5-methyl (4R)-2,6-dimethyl-4-(3-nitrophenyl)-1,4-dihydropyridine-3,5-dicarboxylate chloride
Synonyms: hydrogen benidipine chloride
Molecular Weight (g/mol): 542.03
Molecular Formula: C28H32ClN3O6
InChi Key: KILKDKRQBYMKQX-MIPPOABVSA-N
SMILES: [H+].[Cl-].COC(=O)C1=C(C)NC(C)=C([C@@H]1C1=CC=CC(=C1)[N+]([O-])=O)C(=O)O[C@@H]1CCCN(CC2=CC=CC=C2)C1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)