accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NNSOOCl

CAS RN 916766-81-9

IUPAC Name: 4-(1-methyl-1H-pyrazol-3-yl)benzene-1-sulfonyl chloride
Synonyms: 4-(1-methylpyrazol-3-yl)benzenesulfonyl chloride
Molecular Weight (g/mol): 256.7
Molecular Formula: C10H9ClN2O2S
InChi Key: PXFXKTMOZHYJJU-UHFFFAOYSA-N
SMILES: CN1C=CC(=N1)C1=CC=C(C=C1)S(Cl)(=O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results