CAS RN 916766-82-0
IUPAC Name:
4-(1-methyl-1H-pyrazol-3-yl)aniline
Synonyms:
4-(1-methylpyrazol-3-yl)aniline
Molecular Weight (g/mol):
173.22
Molecular Formula:
C10H11N3
InChi Key:
WEFJHPBVOKVCLZ-UHFFFAOYSA-N
SMILES:
CN1C=CC(=N1)C1=CC=C(N)C=C1
1
–
1
of
1
results
1
4-(1-Methyl-1H-pyrazol-3-yl)aniline, 97%, Thermo Scientific™
CAS: 916766-82-0 Molecular Formula: C10H11N3 Molecular Weight (g/mol): 173.219 MDL Number: MFCD03407387 InChI Key: WEFJHPBVOKVCLZ-UHFFFAOYSA-N PubChem CID: 18526375 IUPAC Name: 4-(1-methylpyrazol-3-yl)aniline SMILES: CN1C=CC(=N1)C2=CC=C(C=C2)N