accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 91678-81-8

IUPAC Name: 3,5-dimethylpyrazin-2-amine
Synonyms: 3,5-dimethylpyrazin-2-amine
Molecular Weight (g/mol): 123.16
Molecular Formula: C6H9N3
InChi Key: GZZRWBRYYZGTNW-UHFFFAOYSA-N
SMILES: CC1=CN=C(N)C(C)=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results