accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 916790-87-9

IUPAC Name: methyl({[4-(thiophen-2-yl)oxan-4-yl]methyl})amine
Synonyms: methyl({[4-(thiophen-2-yl)oxan-4-yl]methyl})amine
Molecular Weight (g/mol): 211.32
Molecular Formula: C11H17NOS
InChi Key: DBLNZQVTWSJQDY-UHFFFAOYSA-N
SMILES: CNCC1(CCOCC1)C1=CC=CS1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results