accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 916792-14-8

ONO

CAS RN 916792-14-8

IUPAC Name: 6-(cyclopentyloxy)pyridine-3-carbaldehyde
Synonyms: 6-(cyclopentyloxy)pyridine-3-carbaldehyde
Molecular Weight (g/mol): 191.23
Molecular Formula: C11H13NO2
InChi Key: NQJPWJIUEZZENP-UHFFFAOYSA-N
SMILES: O=CC1=CN=C(OC2CCCC2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results