accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 91692-15-8

IUPAC Name: 4-chloro-N,N-bis(2-methylpropyl)butanamide
Synonyms: 4-chloro-N,N-bis(2-methylpropyl)butanamide
Molecular Weight (g/mol): 233.78
Molecular Formula: C12H24ClNO
InChi Key: ZAOYOLSPPPCZNR-UHFFFAOYSA-N
SMILES: CC(C)CN(CC(C)C)C(=O)CCCCl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results