accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 917-61-3

IUPAC Name: sodium cyanate
Synonyms: sodium cyanate
Molecular Weight (g/mol): 65.01
Molecular Formula: CNNaO
InChi Key: ZVCDLGYNFYZZOK-UHFFFAOYSA-M
SMILES: [Na+].[O-]C#N
Molecular Weight (g/mol) 
  • (3)
  • (1)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
Percent Purity 
  • (1)
  • (3)
Grade 
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (4)
  • (3)
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)
  • (1)
  • (1)