accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 91714-93-1

IUPAC Name: sodium 2-[2-amino-3-(4-bromobenzoyl)phenyl]acetate
Synonyms: bromfenac sodium salt
Molecular Weight (g/mol): 356.15
Molecular Formula: C15H11BrNNaO3
InChi Key: HZFGMQJYAFHESD-UHFFFAOYSA-M
SMILES: [Na+].NC1=C(CC([O-])=O)C=CC=C1C(=O)C1=CC=C(Br)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results