accessibility menu, dialog, popup

UserName

Viewing profile as user:
FFFSOONSOOFFFCs

CAS RN 91742-16-4

IUPAC Name: caesium(1+) trifluoro[(trifluoromethanesulfonylazanidyl)sulfonyl]methane
Synonyms: caesium(1+) bistriflylimide anion
Molecular Weight (g/mol): 413.04
Molecular Formula: C2CsF6NO4S2
InChi Key: NDUUPYVMAUBBMO-UHFFFAOYSA-N
SMILES: [Cs+].FC(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results