accessibility menu, dialog, popup

UserName

Viewing profile as user:
OONH2

CAS RN 917483-71-7

IUPAC Name: 4-(oxan-4-yloxy)aniline
Synonyms: 4-(oxan-4-yloxy)aniline
Molecular Weight (g/mol): 193.25
Molecular Formula: C11H15NO2
InChi Key: CCIFXNITZHWCRR-UHFFFAOYSA-N
SMILES: NC1=CC=C(OC2CCOCC2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results