accessibility menu, dialog, popup

UserName

Viewing profile as user:
PPPdClClFeCyCyCyCy

CAS RN 917511-90-1

IUPAC Name: Dichloro[1,1'-bis(dicyclohexylphosphino)ferrocene]palladium(II)
Synonyms: —
Molecular Weight (g/mol): 755.9
Molecular Formula: C34H52Cl2FeP2Pd
InChi Key: —
SMILES: [Fe].Cl[Pd]Cl.C1CCC(CC1)P(C1CCCCC1)c1cccc1.C1CCC(CC1)P(C1CCCCC1)c1cccc1
Molecular Weight (g/mol) 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)